Colorimetric and Fluorogenic Enzyme Substrates, Biotium
BCIP/NBT Kit, 1 kit
Catalog # 89138-192
Supplier: Biotium
Supplier #: 10003
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Catalog # 89138-192
Supplier: Biotium
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Popular chromogenic and fluorogenic substrates for phosphatases, peroxidases, and other enzymes.
- Wide selection of popular substrates for alkaline phosphatase, horseradish peroxidase, beta-galactosidase, and other common enzymes
- Chromogenic substrates for absorbance-based detection or light microscopy
- Fluorogenic substrates for highly sensitive fluorescence-based ELISA
- Substrate sampler kits available
Biotium offers a number of popular substrates for alkaline phosphatase, beta-galactosidase, and horseradish peroxidase which are widely used for chromogenic or fluorogenic ELISA, immunohistochemistry, or blotting applications.
Beta-glucuronidase substrates are widely used to detect E. coli bacterial contamination in food and water, and also in reporter gene assays in plants.
X-beta-D-cellobioside is a substrate for beta-cellobiosidase.
ABTS is a registered trademark of Roche. Amplex Red is a registered trademark of Thermo Fisher Scientific.
Specifications
Specifications
- Désignation:BCIP/NBT Kit
- Taille:1 kit