Miscellaneous Dissolution Accessories, Quality Lab Accessories
Short Replacement Needle for VK8000 Auto-Sampler
Catalog # 89133-044
Supplier: QLA
: REPNED-VS
Miscellaneous Dissolution Accessories, Quality Lab Accessories
Catalog # 89133-044
Supplier: QLA
Jump to:
Product Details & Documents
Everclear Water Bath Preservative is a crystalline formula offered in premeasured vials for use with dissolution water baths.Use with DEIONIZED water
The following products are compatible with Agilent/VanKel equipment: APP7 accessories, Rotating Bottle accessories, Auto-sampler accessories and Paper Rolls.
PTFE Suspension cups can be used with most dissolution baths.
A single dose will protect a bath for up to 8 weeks.
Specifications
- Désignation:Short Replacement Needle for VK8000 Auto-Sampler