Sampling System, Sterile, Advanced Scientifics
4× 1000 ml (33.8 oz.)
Catalog # 89127-380
Supplier: Advanced Scientific, Inc.
Supplier #: 4MP0038
Sampling System, Sterile, Advanced Scientifics
Catalog # 89127-380
Supplier: Advanced Scientific, Inc.
Jump to:
Product Details & Documents
Polyethylene sample bags complete with 30.5 cm (12") c-flex tubing, pinch and cut devices, male luer injection sites, and 3.8 cm (1½") sanitary sampling system valve.
Specifications
- Capacité:4× 1000 ml (33.8 oz.)