Sampling System, Sterile, Advanced Scientifics
Sampling System, Sterile, Advanced Scientifics
4× 1000 ml (33.8 oz.)
Catalog # 89127-380
Supplier #:  4MP0038
Sampling System, Sterile, Advanced Scientifics
Catalog # 89127-380
Sampling System, Sterile, Advanced Scientifics
Sampling System, Sterile, Advanced Scientifics
4× 1000 ml (33.8 oz.)
Price
$2,602.76
Log in for availability
Case of 8
Product Details & Documents
Polyethylene sample bags complete with 30.5 cm (12") c-flex tubing, pinch and cut devices, male luer injection sites, and 3.8 cm (1½") sanitary sampling system valve.
Specifications
  • Capacité:
    4× 1000 ml (33.8 oz.)