Sampling System, Sterile, Advanced Scientifics | Avantor
Sampling System, Sterile, Advanced Scientifics
4× 500 ml (16.9 oz.)
Catalog # 89127-378
Fournisseur #:  4MP0037
Sampling System, Sterile, Advanced Scientifics
Catalog # 89127-378
Sampling System, Sterile, Advanced Scientifics
Sampling System, Sterile, Advanced Scientifics
4× 500 ml (16.9 oz.)
Price
2 602,76 $
Log in pour la disponibilité
Case of 8
Product Details & Documents
Polyethylene sample bags complete with 30.5 cm (12") c-flex tubing, pinch and cut devices, male luer injection sites, and 3.8 cm (1½") sanitary sampling system valve.
Spécifications
  • Capacité:
    4× 500 ml (16.9 oz.)