Sampling System, Sterile, Advanced Scientifics
4× 500 ml (16.9 oz.)
Catalog # 89127-378
Supplier: Advanced Scientific, Inc.
Fournisseur #: 4MP0037
Sampling System, Sterile, Advanced Scientifics
Catalog # 89127-378
Supplier: Advanced Scientific, Inc.
Jump to:
Product Details & Documents
Polyethylene sample bags complete with 30.5 cm (12") c-flex tubing, pinch and cut devices, male luer injection sites, and 3.8 cm (1½") sanitary sampling system valve.
Spécifications
- Capacité:4× 500 ml (16.9 oz.)