VWR® Labels 'Hazard Warning'
Warning label, Flammable, Yellow on red, 102×13 mm (4×0.5")
89097-948
Distinctive labels warn workers of hazardous conditions or materials.
- Easy to see
- Distinctive colors
: Supplied 125 labels per roll, 1 roll per case.
- Color:Yellow on red
- Désignation:Warning label
- Dimensions:102×13 mm (4×0.5")
- Legend:Flammable
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



