VWR® Labels 'Hazard Warning'
Warning label, Contaminated, Black on fluorescent green, 102×13 mm (4×0.5")
Catalog # 89097-944
Supplier: VWR International
Jump to:
Product Details & Documents
Distinctive labels warn workers of hazardous conditions or materials.
- Easy to see
- Distinctive colors
Ordering information: Supplied 125 labels per roll, 1 roll per case.
Specifications
- Color:Black on fluorescent green
- Désignation:Warning label
- Dimensions:102×13 mm (4×0.5")
- Legend:Contaminated



