VWR® Protective Labels
Write-on hazard warning labels, 38.1×76.2 mm (1.5×3")
Catalog # 89097-894
Supplier: VWR International
Jump to:
Product Details & Documents
Radiant orange write-on labels with a protective cover prevent the loss of valuable information.
- No dispenser required
- Easy and simple application
Designed for labeling reagents, samples, or noting calibration data.
Labels protect written information from tampering, moisture, smearing, heat and cold, chemicals, and solvents.
Simply write information on label, peel backing from transparent film, and press film over the label. Apply the label to any clean, dry surface.
Packaging: 500 labels per roll.
Specifications
- Désignation:Write-on hazard warning labels
- Dimensions:38.1×76.2 mm (1.5×3")



