VWR® Laser Printer Labels | Avantor
VWR® Laser Printer Labels
Printer labels for laser printers, 19.1×67 mm (0.75×2.64"), 42 labels/sheet, 100 sheets (4200 labels)
Catalog # 89097-866
Supplier:   VWR International
VWR® Laser Printer Labels
Catalog # 89097-866
Supplier:  VWR International
VWR® Laser Printer Labels
VWR® Laser Printer Labels
Printer labels for laser printers, 19.1×67 mm (0.75×2.64"), 42 labels/sheet, 100 sheets (4200 labels)
Price
$166.64
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 100
Product Details & Documents
Laser printer-compatible labels are designed for use in laboratory or office applications.
  • Supplied in a rainbow of colors
  • Removable or permanent adhesives depending on color
Supplied on 8¹/₂×11" sheets, all labels include step-by-step instructions on how to create, format, and print individual labels.
Fluorescent rainbow labels are supplied in sets of 25 laser sheets, with 5 each of fluorescent green, orange, chartreuse, pink, and red. The labels feature removeable adhesive, with the exception of fluorescent red, which is manufactured with a permanent adhesive. The white single-ply labels feature a permanent adhesive.
Spécifications
  • Désignation:
    Printer labels for laser printers
  • Dimensions:
    19.1×67 mm (0.75×2.64")
  • Packaging:
    42 labels/sheet, 100 sheets (4200 labels)