VWR® Stat Labels
Stat labels, 12.7×25.4 mm (0.5×1"), 1000 labels/roll, 1 roll
89097-864
Labels feature a bold black print atop a fluorescent red background.
- Permanent adhesive
- Highly visible label
: Each case contains one roll of 1000 labels.
- Désignation:Stat labels
- Dimensions:12.7×25.4 mm (0.5×1")
- Packaging:1000 labels/roll, 1 roll



