Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Slide Labels | Avantor

VWR® Slide Labels

Slide labels, White, 22.2×22.2 mm (0.88×0.88"), 40 labels/sheet, 25 sheets (100 labels)
Catalog # 89097-842
Supplier:   VWR International
VWR® Slide Labels
Catalog # 89097-842
Supplier:  VWR International
VWR® Slide Labels
VWR® Slide Labels

Slide labels, White, 22.2×22.2 mm (0.88×0.88"), 40 labels/sheet, 25 sheets (100 labels)

Price
$38.74
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 1
Product Details & Documents

Designed to help prevent curling or detaching, slide labels are printed on smudgeproof litho sheets and feature a permanent adhesive.

  • Labels have a thickness 0.012 cm (0.0078")
  • Color: white
Spécifications
  • Color:
    White
  • Désignation:
    Slide labels
  • Dimensions:
    22.2×22.2 mm (0.88×0.88")
  • Packaging:
    40 labels/sheet, 25 sheets (100 labels)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.