VWR® Labeling Dots, Self-Adhesive
Labeling dots, self-adhesive, White, 1.9 cm (3/4")
Supplier: VWR International
Jump to:
Product Details & Documents
Designed for use on test tubes, sample batches, or virtually any surface that requires labeling.
- Hundreds of uses
- Dots available in six colors
Labels can also be used for date coding or personal identification. Labeling dots are packaged 1000 per roll, 1 roll per case.
Spécifications
- Color:White
- Désignation:Labeling dots, self-adhesive
- Diameter:1.9 cm (3/4")