Biohazard Wipes and Liners, Current Technologies
Heavy Absorbency Biohazard Wipes, 40.6 x 40.6 cm (16 x 16")
  89092-534
 :  BH 10600
Biohazard Wipes and Liners, Current Technologies
  89092-534
Biohazard Wipes and Liners, Current Technologies
Biohazard Wipes and Liners, Current Technologies
Heavy Absorbency Biohazard Wipes, 40.6 x 40.6 cm (16 x 16")
$565.58
Case of 2
Low-linting wipes and liners are specifically designed to protect workers and labs from contamination from blood, body fluids, tissue, specimens plus chemicals.
Wipes and liners are available in several sizes and in large rolls for cutting to size. Wipes are also available in standard absorbency, which can absorb 9× its own weight, heavy absorbency, which can absorb 12× its weight and super absorbency, which can absorb 25× its weight.
Specifically sized dispensers are constructed of clear acrylic.
Absorbent white side soaks up blood, tissue, specimen, body fluids, and chemicals. Bright orange barrier side, made of coextruded plastic, blocks soak-through to gloves, hands and counters, limiting the spread of contamination. Wipes are impermeable to strong solvents including xylene and xylene substitutes, acetone, ketones, alcohols, and formalin. Use to pop caps, wipe pipets and probes, handle specimens and line benches, counters and trays, clean up spills.
 : Bio-Screen wipes and liners are non-sterile and are not for use on patients.
  • Désignation:
    Heavy Absorbency Biohazard Wipes
  • Dimensions:
    40.6 x 40.6 cm (16 x 16")
Accessories
  •  
    Description
    Material
  • 16200-180
    Dispenser for 10.2 x 10.2 cm (4 x 4") Wipes/Towels
    Acrylic
    Each - $97.22
  •  
    Description
    Material
  • 16003-778
    Dispenser for 7.6 x 7.6 cm (3 x 3") Wipes/Towels
    Acrylic
    Each - $97.65