PYREX® Reducing Adapters with [ST] Joints, Corning | Avantor
PYREX® Reducing Adapters with [ST] Joints, Corning
105mm, 24/40, [ST] 29/42
Catalog # 89091-644
   Low Manufacturing Impact
Supplier:   Corning
Fournisseur #:  8800-2924
PYREX® Reducing Adapters with [ST] Joints, Corning
Catalog # 89091-644
Supplier:  Corning
   Low Manufacturing Impact
PYREX® Reducing Adapters with [ST] Joints, Corning
PYREX® Reducing Adapters with [ST] Joints, Corning
105mm, 24/40, [ST] 29/42
Price
876,00 $
Log in pour la disponibilité
Case of 6
Product Details & Documents
PYREX® glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
Spécifications
  • Longueur:
    105mm
  • Bottom Inner [ST] Joint Size:
    24/40
  • Top Outer ST Joint Size:
    [ST] 29/42
  • Cat. No.:
    89091-644
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact