PYREX® Reducing Adapters with [ST] Joints, Corning
105mm, 24/40, [ST] 29/42
Catalog # 89091-644
Supplier: Corning
Fournisseur #: 8800-2924
PYREX® Reducing Adapters with [ST] Joints, Corning
Catalog # 89091-644
Supplier: Corning
Jump to:
Product Details & Documents
PYREX® glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
Spécifications
- Longueur:105mm
- Bottom Inner [ST] Joint Size:24/40
- Top Outer ST Joint Size:[ST] 29/42
- Cat. No.:89091-644
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact