PYREX® Reducing Adapters with [ST] Joints, Corning
105 mm, 19/38, [ST] 24/40
Catalog # 89091-642
Product available on GSA Advantage®
Supplier: Corning
Supplier #: 8800-2419
PYREX® Reducing Adapters with [ST] Joints, Corning
Catalog # 89091-642
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
PYREX® glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
Spécifications
- Longueur:105 mm
- Bottom Inner [ST] Joint Size:19/38
- Top Outer ST Joint Size:[ST] 24/40
- Cat. No.:89091-642
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact