PYREX® Serum Bottles, Corning
PYREX® Serum Bottles, Corning
9 L (2.4 gal.)
Catalog # 89091-534
Supplier:   Corning
Supplier #:  1585-9L
PYREX® Serum Bottles, Corning
Catalog # 89091-534
Supplier:  Corning
PYREX® Serum Bottles, Corning
PYREX® Serum Bottles, Corning
9 L (2.4 gal.)
Price
$1,800.00
Log in pour la disponibilité
Case of 4
Product Details & Documents
Designed for the handling and storage of sterile culture media and sera when the stability of glass is of prime importance.
  • Fits a #8 rubber stopper
  • Stopper not included
  • Bottle withstands repeated wet or dry autoclaving
These serum bottles are made of Pyrex® borosilicate glass.
Also suitable for storing distilled water and standard solutions. Necks are tooled for uniform rubber stopper fit and increased mechanical strength.
Spécifications
  • Capacité:
    9 L (2.4 gal.)