Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning | Avantor

PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning

5 L, 11, 221 x 310 mm (811/16 x 123/16")
Catalog # 89091-474
Supplier:   Corning
Fournisseur #:  4260-5L
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
Catalog # 89091-474
Supplier:  Corning
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning

5 L, 11, 221 x 310 mm (811/16 x 123/16")

Price
$1,920.00
Log in pour la disponibilité
Case of 6
Product Details & Documents

PYREX® borosilicate glass, clear, with short neck.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

These flask mouth tooled for uniform rubber stopper fit. The normal working temperature is 230 °C.

Spécifications
  • Capacité:
    5 L
  • Stopper No.:
    11
  • O.D.×H:
    221 x 310 mm (811/16 x 123/16")