PYREX® Serum Bottles, Corning
4 L (1 gal.)
Supplier: Corning
Fournisseur #: 1585-4L
Jump to:
Product Details & Documents
Designed for the handling and storage of sterile culture media and sera when the stability of glass is of prime importance.
- Fits a #8 rubber stopper
- Stopper not included
- Bottle withstands repeated wet or dry autoclaving
These serum bottles are made of Pyrex® borosilicate glass.
Also suitable for storing distilled water and standard solutions. Necks are tooled for uniform rubber stopper fit and increased mechanical strength.
Spécifications
- Capacité:4 L (1 gal.)