Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Corning® Solid Black and White Polystyrene Microplates, 384 Well

Corning® Solid Black and White Polystyrene Microplates, 384 Well

Corning® Low Volume 384 Well Black Flat Bottom Polystyrene TC-Treated Microplate, With Lid, 50 µl, 50 µl, 5 - 40 µl, Black, Sterile, Tissue culture (TC), Flat-Bottom, 10/Bag
Catalog # 89089-860
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Supplier #:  3822
Corning® Solid Black and White Polystyrene Microplates, 384 Well
Catalog # 89089-860
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Corning® Solid Black and White Polystyrene Microplates, 384 Well
Corning® Solid Black and White Polystyrene Microplates, 384 Well

Corning® Low Volume 384 Well Black Flat Bottom Polystyrene TC-Treated Microplate, With Lid, 50 µl, 50 µl, 5 - 40 µl, Black, Sterile, Tissue culture (TC), Flat-Bottom, 10/Bag

Price
$1,096.50
New Customer Promotional Price
Log in for availability
Case of 50
Product Details & Documents

PS. Designed to reduce well-to-well crosstalk during fluorescent and luminescent assays.

  • Square well top for easier robotic pipetting
  • Round bottom for better Z factor and minimized trapped air
  • Conical well molded in the shape of a light cone for efficiency
  • Black walls reduce well-to-well crosstalk and autofluorescence during fluorescent assays

High binding surface is a general purpose surface that binds medium (>10kD) and large biomolecules possessing ionic groups and/or hydrophobic regions. Binding capacity is approximately 400 to 500ng IgG/cm.

Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions. Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm²).

Nonbinding surface (NBS™) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise.

Specifications
  • Sterility:
    Sterile
  • Well Volume:
    50 µl
  • Color:
    Black
  • Désignation:
    Corning® Low Volume 384 Well Black Flat Bottom Polystyrene TC-Treated Microplate, With Lid
  • Packaging:
    10/Bag
  • Well Shape:
    Flat-Bottom
  • Surface Treatment:
    Tissue culture (TC)
  • Working Well Volume:
    5 - 40 µl
  • Total Well Volume:
    50 µl
  • Cat. No.:
    89089-860
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact