Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Corning® Disposable Spinner Flasks, Corning | Avantor

Corning® Disposable Spinner Flasks, Corning

Corning® 125 ml Disposable Spinner Flask, 125 ml, 25 mm, 63.5×145 mm, Plain
Catalog # 89089-720
Supplier:   Corning
Fournisseur #:  3152
Corning® Disposable Spinner Flasks, Corning
Catalog # 89089-720
Supplier:  Corning
Corning® Disposable Spinner Flasks, Corning
Corning® Disposable Spinner Flasks, Corning

Corning® 125 ml Disposable Spinner Flask, 125 ml, 25 mm, 63.5×145 mm, Plain

Price
$2,244.00
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 12
Product Details & Documents

The Corning® disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.

  • Paddle size and height is optimised for each vessel size
  • Integrated magnet provides smooth, even rotation at required speeds on slow-speed stirrers
  • Heat build-up in the vessel is reduced by means of a specially designed flange that raised the vessel off the stir-plate surface for the 125 and 500 ml flasks only

Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.

Made of USP Class VI and ISO 10993-18 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.

Spécifications
  • Capacité:
    125 ml
  • Désignation:
    Corning® 125 ml Disposable Spinner Flask
  • Style:
    Plain
  • Diameter ×H:
    63.5×145 mm
  • Sidearm Neck O.D.:
    25 mm
Accessories

Informations sur le produit

  • Catalog #
    Material
    Sterility
    Availability
    Price
  • Vent cap
    Vent cap
    89089-914
    High Density Polyethylene, orange
    Sterile
    Log in pour la disponibilité
    Case of 4 - 68,85 $

Informations sur le produit

  • Catalog #
    Material
    Sterility
    Availability
    Price
  • Aseptic transfer cap
    Aseptic transfer cap
    89089-910
    High Density Polyethylene, orange
    Sterile
    Log in pour la disponibilité
    Case of 2 - 630,70 $

Informations sur le produit

  • Catalog #
    Fits Flask
    Availability
    Price
  • Modified Impeller Assembly for Magnetically-Driven 36L Corning® Proculture® Spinner Flask, Reusable
    Modified Impeller Assembly for Magnetically-Driven 36L Corning® Proculture® Spinner Flask, Reusable
    89089-950
    36L Corning® Proculture® spinner flask (22877-088, 22877-098, 22877-108, 22877-132, 22877-138, 22877-142)
    Log in pour la disponibilité
    Case of 1 - 1 450,00 $

Informations sur le produit

  • Catalog #
    Material
    Sterility
    Availability
    Price
  • Aseptic transfer cap
    Aseptic transfer cap
    89089-912
    High Density Polyethylene, orange
    Sterile
    Log in pour la disponibilité
    Case of 2 - 689,35 $

Informations sur le produit

  • Catalog #
    Material
    Sterility
    Availability
    Price
  • Aseptic transfer cap
    Aseptic transfer cap
    89089-908
    High Density Polyethylene, orange
    Sterile
    Log in pour la disponibilité
    Case of 2 - 641,75 $

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.