Coliform Sampling Bottles, I-CHEM
Polypropylene bottle, with 50 mg tablet, 120 ml (4 oz.), 38-400 mm
  89088-064
 :  P156-120PP
Coliform Sampling Bottles, I-CHEM
  89088-064
Coliform Sampling Bottles, I-CHEM
Coliform Sampling Bottles, I-CHEM
Polypropylene bottle, with 50 mg tablet, 120 ml (4 oz.), 38-400 mm
1 011,82 $
Case of 200
Designed for use with EPA compliant total coliform and fecal coliform water analysis using the Colilert analysis method or equivalent.
These screw-top sterile coliform water sample bottles are available in a variety of sizes and materials, with or without sodium thiosulfate for sample dechlorination.
  • Taille du bouchon:
    38-400 mm
  • Capacité:
    120 ml (4 oz.)
  • Désignation:
    Polypropylene bottle, with 50 mg tablet