Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Coliform Sampling Bottles, I-CHEM

Coliform Sampling Bottles, I-CHEM

Polyethylene bottle, without tablet, 250 ml (8.5 oz.), 45-400 mm
Catalog # 89088-062
Supplier:   Thermo Scientific
Supplier #:  156-250PE
Coliform Sampling Bottles, I-CHEM
Catalog # 89088-062
Supplier:  Thermo Scientific
Coliform Sampling Bottles, I-CHEM
Coliform Sampling Bottles, I-CHEM

Polyethylene bottle, without tablet, 250 ml (8.5 oz.), 45-400 mm

Price
2 515,64 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 228
Product Details & Documents

Designed for use with EPA compliant total coliform and fecal coliform water analysis using the Colilert analysis method or equivalent.

These screw-top sterile coliform water sample bottles are available in a variety of sizes and materials, with or without sodium thiosulfate for sample dechlorination.

Spécifications
  • Taille du bouchon:
    45-400 mm
  • Capacité:
    250 ml (8.5 oz.)
  • Désignation:
    Polyethylene bottle, without tablet
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.