PTFE Plug Inserts, Ace Glass
PTFE Plug Inserts, Solid Plug, 1/4" NPT
Catalog # 89084-118
Supplier: Ace Glass
Fournisseur #: 12867-04
Jump to:
Product Details & Documents
These are PTFE inserts for PTFE reaction vessel heads.
Solid plug.
Spécifications
- Désignation:PTFE Plug Inserts, Solid Plug
- Taille:1/4" NPT