Miscellaneous Dissolution Accessories, Quality Lab Accessories
100 mL Clear Glass Bottle and Decant Cap with 40 Mesh SS Screen
Catalog # 89080-338
Supplier: QLA
Fournisseur #: RBAASSY-02
Miscellaneous Dissolution Accessories, Quality Lab Accessories
Catalog # 89080-338
Supplier: QLA
Jump to:
Product Details & Documents
Everclear Water Bath Preservative is a crystalline formula offered in premeasured vials for use with dissolution water baths.Use with DEIONIZED water
The following products are compatible with Agilent/VanKel equipment: APP7 accessories, Rotating Bottle accessories, Auto-sampler accessories and Paper Rolls.
PTFE Suspension cups can be used with most dissolution baths.
A single dose will protect a bath for up to 8 weeks.
Spécifications
- Désignation:100 mL Clear Glass Bottle and Decant Cap with 40 Mesh SS Screen