Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Condenser, Allihn

Condenser, Allihn

Condenser, Allihn, 24/25, 200 mm
Catalog # 89053-808
Supplier:   Ace Glass
Supplier #:  5945-34
Condenser, Allihn
Catalog # 89053-808
Supplier:  Ace Glass
Condenser, Allihn
Condenser, Allihn

Condenser, Allihn, 24/25, 200 mm

Price
$410.81
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

Bulb type Allihn condenser, approximately one bulb per every 50 mm, with standard taper inner joint at bottom and reinforced standard taper outer joint at top

Fabricated from borosilicate glass. Size D glass hose connections, for use with 3/8-inch I.D. tubing.

Spécifications
  • ST Joint:
    24/25
  • Désignation:
    Condenser, Allihn
  • Jacket Length:
    200 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.