Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Condenser, Allihn, Jacketed | Avantor

Condenser, Allihn, Jacketed

Condenser, Alihn, Jacketed, 300 mm
Catalog # 89053-792
Supplier:   Ace Glass
Fournisseur #:  5941-14
Condenser, Allihn, Jacketed
Catalog # 89053-792
Supplier:  Ace Glass
Condenser, Allihn, Jacketed
Condenser, Allihn, Jacketed

Condenser, Alihn, Jacketed, 300 mm

Price
$414.78
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

Bulb type Allihn condenser, approximately one bulb per every 50 mm, with 24/40 standard taper inner joint at bottom, open tube at the top

Fabricated from borosilicate glass. Use with 3/8-inch I.D. tubing, size D hose connections.

Spécifications
  • Désignation:
    Condenser, Alihn, Jacketed
  • Jacket Length:
    300 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.