Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Test Tubes, Greiner Bio-One

Test Tubes, Greiner Bio-One

15 ml, With Blue Screw Cap, 120 mm, 17 mm
Catalog # 89049-098
Supplier #:  186171
Test Tubes, Greiner Bio-One
Catalog # 89049-098
Supplier:  
Test Tubes, Greiner Bio-One
Test Tubes, Greiner Bio-One

15 ml, With Blue Screw Cap, 120 mm, 17 mm

Discontinued
Product Details & Documents

These high-quality test tubes are available in a wide variety of materials, sizes, and volume ranges.

  • Excellent optical clarity
  • Round, flat, or conical bottom designs available
  • Polycarbonate, polypropylene, or polystyrene construction available
  • Exceptional thermal and chemical stability

Test tubes provide excellent optical clarity, and are available in round or conical bottom designs. Unless otherwise noted, tube caps are not included.

 : Test tube 82050-248 is supplied sterile.

Spécifications
  • Ø:
    17 mm
  • Capacité:
    15 ml
  • Désignation:
    With Blue Screw Cap
  • Hauteur:
    120 mm
Accessories

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Screw Cap for Polypropylene Tube, White, For Model 163201
    Screw Cap for Polypropylene Tube, White, For Model 163201
    10066-772
    Log in pour la disponibilité
    Case of 9,000 - $935.15