Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Buchner Funnels, Table-Type

VWR® Buchner Funnels, Table-Type

750 ml, 109 mm
Catalog # 89038-112
Supplier:   VWR International
VWR® Buchner Funnels, Table-Type
Catalog # 89038-112
Supplier:  VWR International
VWR® Buchner Funnels, Table-Type
VWR® Buchner Funnels, Table-Type

750 ml, 109 mm

Price
1 096,60 $
New Customer Promotional Price
Log in for availability
Case of 1
Product Details & Documents

These chemical-porcelain funnels are ideal for vacuum filtration and feature a slanting bottom that leads to a tubulated side outlet.

  • Perforated plate
  • Autoclavable

Designed with a wide mouth for convenience and a wide base that rests firmly on a tabletop for stability.

Specifications
  • Capacité:
    750 ml
  • Hauteur:
    109 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.