Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
50 L, Insulating Jacket
Catalog # 89029-978
Supplier:   Chemglass
Supplier #:  CG-10011-151
Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
Catalog # 89029-978
Supplier:  Chemglass
Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
50 L, Insulating Jacket
Price
$6,987.58
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Insulating jacket is made from a heavy fiberglass fabric on the interior with a silicone-coated fiberglass fabric on the exterior.
  • Jacket is slit lengthwise to a hole in the bottom center
  • With lacing closure to accommodate a bottom drain valve
For use as a heat or cold shield on chemglass jacketed reactors.
Specifications
  • Capacité:
    50 L
  • Désignation:
    Insulating Jacket