Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
50 L, Insulating Jacket
Catalog # 89029-978
Supplier: Chemglass
Supplier #: CG-10011-151
Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
Catalog # 89029-978
Supplier: Chemglass
Jump to:
Product Details & Documents
Insulating jacket is made from a heavy fiberglass fabric on the interior with a silicone-coated fiberglass fabric on the exterior.
- Jacket is slit lengthwise to a hole in the bottom center
- With lacing closure to accommodate a bottom drain valve
For use as a heat or cold shield on chemglass jacketed reactors.
Specifications
- Capacité:50 L
- Désignation:Insulating Jacket