Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
15 L, Insulating Jacket
Catalog # 89029-972
Supplier: Chemglass
Fournisseur #: CG-10011-115
Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
Catalog # 89029-972
Supplier: Chemglass
Jump to:
Product Details & Documents
Insulating jacket is made from a heavy fiberglass fabric on the interior with a silicone-coated fiberglass fabric on the exterior.
- Jacket is slit lengthwise to a hole in the bottom center
- With lacing closure to accommodate a bottom drain valve
For use as a heat or cold shield on chemglass jacketed reactors.
Spécifications
- Capacité:15 L
- Désignation:Insulating Jacket
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.