VWR® Vials, PTFE
50.00 ml, 43×78 mm, Oui
89026-432
PTFE vials are ideal for storing aggressive or valuable materials.
Suitable for small-scale reactions at low pressure. Vials feature an inner taper for easy removal of contents and thick wall construction for durability.
- Avec bouchon:Oui
- Capacité:50.00 ml
- Diameter ×H:43×78 mm