VWR® Vials, PTFE
25.00 ml, 33×64 mm, Oui
Catalog # 89026-430
Supplier: VWR International
Jump to:
Product Details & Documents
PTFE vials are ideal for storing aggressive or valuable materials.
Suitable for small-scale reactions at low pressure. Vials feature an inner taper for easy removal of contents and thick wall construction for durability.
Spécifications
- Avec bouchon:Oui
- Capacité:25.00 ml
- Diameter ×H:33×64 mm