Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Dropping Bottles, PTFE | Avantor

VWR® Dropping Bottles, PTFE

50 ml (1.7 oz.), 43×100 mm
Catalog # 89026-088
Supplier:   VWR International
VWR® Dropping Bottles, PTFE
Catalog # 89026-088
Supplier:  VWR International
VWR® Dropping Bottles, PTFE
VWR® Dropping Bottles, PTFE

50 ml (1.7 oz.), 43×100 mm

Price
351,14 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

PTFE dropping bottles are suitable for dispensing and dropping small volumes of reagents and aggressive chemicals.

  • Chemically resistant and inert
  • Tight, leak-free seal
  • Bottle sides are flexible

Bottle sides are flexible to enable user to squeeze out contents. Screw-on cap ensures a tight, leak-free seal.

Spécifications
  • Capacité:
    50 ml (1.7 oz.)
  • Diameter ×H:
    43×100 mm