Precision Suture Scissors, Delicate, Sklar | Avantor
Precision Suture Scissors, Delicate, Sklar
114 mm (4½"), Curved
Catalog # 89025-320
Supplier:   Sklar
Fournisseur #:  22-2825
Precision Suture Scissors, Delicate, Sklar
Catalog # 89025-320
Supplier:  Sklar
Precision Suture Scissors, Delicate, Sklar
Precision Suture Scissors, Delicate, Sklar
114 mm (4½"), Curved
Price
$283.24
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sharp/sharp blades
Upper blade has fine serrations. Suturing notch. Premium-grade stainless steel. Autoclavable.
Spécifications
  • Longueur:
    114 mm (4½")
  • Blade Type:
    Curved