Precision Suture Scissors, Delicate, Sklar
89 mm (3½"), Straight
Catalog # 89025-314
Supplier:   Sklar
 :  22-2820
Precision Suture Scissors, Delicate, Sklar
Catalog # 89025-314
Supplier:  Sklar
Precision Suture Scissors, Delicate, Sklar
Precision Suture Scissors, Delicate, Sklar
89 mm (3½"), Straight
Price
$296.65
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sharp/sharp blades
Upper blade has fine serrations. Suturing notch. Premium-grade stainless steel. Autoclavable.
Spécifications
  • Longueur:
    89 mm (3½")
  • Blade Type:
    Straight