Dressing Forceps, Delicate, Sklar
Product Details & Documents
Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds
Censitrac Ready OR Grade Dressing Forceps are industry standard and constructed of stainless steel.
This product is straight with serrated tips.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Specifications
- Désignation:Dressing Forceps, Delicate
- Longueur:140 mm (5½")
- Matière:Stainless steel
- Curvature:Straight
- Tip Style:Serrated
- Grade:Premium OR-Grade
- Sterile:-
- Handle:Thumb