Diversified Biotech CoolSafe™ Systems
CoolSafe System for 1.5 mL Cryo Tubes, For 1.5 mL Cryogenic Tubes
Catalog # 89024-866
Supplier:   Diversified Biotech
 :  CSAF-3050
Diversified Biotech CoolSafe™ Systems
Catalog # 89024-866
Diversified Biotech CoolSafe™ Systems
Diversified Biotech CoolSafe™ Systems
CoolSafe System for 1.5 mL Cryo Tubes, For 1.5 mL Cryogenic Tubes
Price
$1,827.28
Log in pour la disponibilité
Each
Product Details & Documents
Anodized aluminum storage kits are ideal for sustaining cool or warm constant temperatures up to 10 hours during benchtop storage
Each kit includes one polystyrene CoolSafe storage box, one anodized aluminum cooling chamber rack, and two interchangeable cool bricks. Additional aluminum cooling chamber racks are also available.
Spécifications
  • Capacité:
    For 1.5 mL Cryogenic Tubes
  • Désignation:
    CoolSafe System for 1.5 mL Cryo Tubes
Accessories
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-880
    Holds Forty-Eight 1.5 mL Tubes
    Log in pour la disponibilité
    Each - $1,301.34
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-882
    Holds Fifty-Five 1.5 mL Cryogenic Tubes
    Log in pour la disponibilité
    Each - $1,289.09
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-884
    Holds One 384-Well Plate, Six 0.5 mL Tubes, and Four 1.5 mL Tubes
    Log in pour la disponibilité
    Each - $2,268.99
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-878
    Holds Ninety-One 0.5 mL Tubes
    Log in pour la disponibilité
    Each - $1,752.18
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-886
    Holds Thirty-Two 5 mL Tubes
    Log in pour la disponibilité
    Each - $1,449.35
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-876
    Holds One PCR Plate or Ninety-Six 0.2 mL Tubes, Six 0.5 mL Tubes, and Four 1.5 mL Tubes
    Log in pour la disponibilité
    Each - $1,462.18
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-888
    Holds Twenty-Four 0.2 mL Tubes, Thirty-Five 0.5 mL Tubes, and Twenty 1.5 mL Tubes
    Log in pour la disponibilité
    Each - $1,396.85
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-890
    Holds One Flat-Bottom Microplate, Six 0.5 mL Tubes, and Four 1.5 mL Tubes
    Log in pour la disponibilité
    Each - $1,326.60