Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Hopper Box, MFG Tray

Hopper Box, MFG Tray

Lid for Box 89022-830, —, White
Catalog # 89022-832
Hopper Box, MFG Tray
Catalog # 89022-832
Supplier:  
Hopper Box, MFG Tray
Hopper Box, MFG Tray

Lid for Box 89022-830, —, White

Discontinued
Product Details & Documents

MFG Tray Hoppers are ideal storage containers for small parts, components, fasteners, and pre-packaged goods

Hopper box lid 89022-832 is designed for use with hopper box 89022-830.

Canted fronts facilitate access to parts, and the stacking design conserves valuable space in the work area. Composite construction ensures strength and dimensional stability, and eliminates corrosion, bending, or end-product contamination.

pH range: 3.0 to 10.0
Temperature range: –51 to 121°C (–60 to 250°F), up to 300°F intermittently

Specifications
  • Color:
    White
  • Désignation:
    Lid for Box 89022-830
  • Dimensions: