Precision Screwdriver, Aven
Phillips Driver, 50 mm (2"), No. 1
Catalog # 89012-008
Fournisseur #: 13913
Jump to:
Discontinued
Product Details & Documents
Specifically designed for electronics
Spécifications
- Désignation:Phillips Driver
- Tip Size:No. 1
- Blade Length:50 mm (2")