Precision Screwdriver, Aven
Phillips Driver, 50 mm (2"), No. 000
Catalog # 89012-002
Fournisseur #: 13910
Jump to:
Discontinued
Product Details & Documents
Specifically designed for electronics
Spécifications
- Désignation:Phillips Driver
- Tip Size:No. 000
- Blade Length:50 mm (2")