Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Precision Screwdriver, Aven

Precision Screwdriver, Aven

Slotted Driver, 50 mm (2"), 3.0 mm
Catalog # 89011-994
Supplier #:  13904
Precision Screwdriver, Aven
Catalog # 89011-994
Supplier:  
Precision Screwdriver, Aven
Precision Screwdriver, Aven

Slotted Driver, 50 mm (2"), 3.0 mm

Discontinued
Product Details & Documents

Specifically designed for electronics

Spécifications
  • Désignation:
    Slotted Driver
  • Tip Size:
    3.0 mm
  • Blade Length:
    50 mm (2")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.