Precision Screwdriver, Aven
Slotted Driver, 50 mm (2"), 1.6 mm
Catalog # 89011-988
Supplier #: 13901
Jump to:
Discontinued
Product Details & Documents
Specifically designed for electronics
Specifications
- Désignation:Slotted Driver
- Tip Size:1.6 mm
- Blade Length:50 mm (2")