VWR® HDPE Multipurpose Containers
VWR® HDPE Multipurpose Containers
5 L (1.3 gal.), 22.3 dia.×17.9H cm (8²⁵/₃₂×7")
Catalog # 89009-674
Supplier:   VWR International
VWR® HDPE Multipurpose Containers
Catalog # 89009-674
Supplier:  VWR International
VWR® HDPE Multipurpose Containers
VWR® HDPE Multipurpose Containers
5 L (1.3 gal.), 22.3 dia.×17.9H cm (8²⁵/₃₂×7")
Price
$940.82
New Customer Promotional Price
Log in for availability
Case of 25
Product Details & Documents
Translucent containers for the storage of pathology, industrial, and environmental samples.
  • HDPE construction
  • Chemical-resistant
  • Translucent siding
Containers are made of high-quality plastic to resist breakage and come in a wide range of sizes. The matching cap snaps on tight for a secure fit. The 4800 mL container is equipped with a handle for portability.
Documents
Specifications
  • Capacité:
    5 L (1.3 gal.)
  • Dimensions:
    22.3 dia.×17.9H cm (8²⁵/₃₂×7")