Torque-Rite® Nuts, Rubber Fab
Torque-Rite® Nut, 50 in/lbs.
Catalog # 89009-464
Jump to:
Discontinued
Product Details & Documents
Torque-Rite® allows you to control compression and expansion while maintaining constant inch/pounds force insuring a perfectsSurface I.D
when used with a Rubber Fab sanitary gasket. Torque-Rite® eliminates the problems associated with over - or under-tightening a gasket which can lead to an unsanitary system.
Spécifications
- Désignation:Torque-Rite® Nut, 50 in/lbs.