Microscope Slide Coverslips, Polysciences | Avantor
Microscope Slide Coverslips, Polysciences
Glass cover slips, rectangular, 40×24 mm, 17 mm
Catalog # 87001-918
Supplier:   Polysciences Inc
Fournisseur #:  24070-1
Microscope Slide Coverslips, Polysciences
Catalog # 87001-918
Supplier:  Polysciences Inc
Microscope Slide Coverslips, Polysciences
Microscope Slide Coverslips, Polysciences
Glass cover slips, rectangular, 40×24 mm, 17 mm
Price
194,59 $
Log in pour la disponibilité
Pack of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rectangular glass coverslips made of Borosilicate glass. Ideal coverslip for various methods, including routine and special stains, immunohistochemistry, and in situ hybridization.
Spécifications
  • Désignation:
    Glass cover slips, rectangular
  • L×l:
    40×24 mm
  • Épaisseur:
    17 mm