Polypropylene Screw Caps, BrandTech | Avantor
Polypropylene Screw Caps, BrandTech
GL18
Catalog # 83009-032
Supplier:   Brandtech
Fournisseur #:  83310
Polypropylene Screw Caps, BrandTech
Catalog # 83009-032
Supplier:  Brandtech
Polypropylene Screw Caps, BrandTech
Polypropylene Screw Caps, BrandTech
GL18
Price
$90.33
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 24
Product Details & Documents
Translucent screw caps are for use with BrandTech polypropylene and low-density polyethylene bottles.
  • Autoclavable translucent polypropylene
  • Fits VITLAB and many other bottles with standard thread
Packaging: Bulk packed.
Spécifications
  • Taille du bouchon:
    GL18