Polypropylene Screw Caps, BrandTech
GL18
Catalog # 83009-032
Supplier: Brandtech
Fournisseur #: 83310
Jump to:
Product Details & Documents
Translucent screw caps are for use with BrandTech polypropylene and low-density polyethylene bottles.
- Autoclavable translucent polypropylene
- Fits VITLAB and many other bottles with standard thread
Packaging: Bulk packed.
Spécifications
- Taille du bouchon:GL18