Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech | Avantor

VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech

50.00 ml, 30×97 mm, Oui
Catalog # 83009-022
Supplier:   Brandtech
Fournisseur #:  69194
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
Catalog # 83009-022
Supplier:  Brandtech
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech

50.00 ml, 30×97 mm, Oui

Price
119,18 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 10
Product Details & Documents

General-purpose sample vials for a variety of lab applications.

  • Polypropylene
  • LDPE push-on 'snap' caps
  • Vials are autoclavable, caps are not

Translucent polypropylene vials include low-density polyethylene push-on caps. Vials and caps are suitable for microwave use.

Spécifications
  • Avec bouchon:
    Oui
  • Capacité:
    50.00 ml
  • Diameter ×H:
    30×97 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.