VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech | Avantor
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
18.00 ml, 22×57 mm, Oui
Catalog # 83009-019
Supplier:   Brandtech
Fournisseur #:  68894
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
Catalog # 83009-019
Supplier:  Brandtech
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
18.00 ml, 22×57 mm, Oui
Price
$255.49
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 25
Product Details & Documents
General-purpose sample vials for a variety of lab applications.
  • Polypropylene
  • LDPE push-on 'snap' caps
  • Vials are autoclavable, caps are not
Translucent polypropylene vials include low-density polyethylene push-on caps. Vials and caps are suitable for microwave use.
Spécifications
  • Avec bouchon:
    Oui
  • Capacité:
    18.00 ml
  • Diameter ×H:
    22×57 mm