Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VITLAB® Forceps, POM, BrandTech | Avantor

VITLAB® Forceps, POM, BrandTech

180 mm (7⅛")
Catalog # 83009-014
Supplier:   Brandtech
Fournisseur #:  68299
VITLAB® Forceps, POM, BrandTech
Catalog # 83009-014
Supplier:  Brandtech
VITLAB® Forceps, POM, BrandTech
VITLAB® Forceps, POM, BrandTech

180 mm (7⅛")

Price
$107.65
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 5
Product Details & Documents

Polyoxymethylene (POM) forceps with rounded ends.

  • Rounded ends
  • Opaque orange-yellow

POM is suitable for autoclaving but not suitable for microwaves. Frequent autoclaving may reduce mechanical stability. Resistant to aliphatic alcohols, ether, aldehydes, aliphatic hydrocarbons, aromatic hydrocarbons, halogenated hydrocarbons, ketones, and alkalis.

Operating temperature: –40 to 130°C (–40 to 266°F)

 : Not recommended for use with ester, acids, and oxidizing agents.

Spécifications
  • Longueur:
    180 mm (7⅛")