VITLAB® Forceps, POM, BrandTech
145 mm (5¾")
  83009-013
 :   Brandtech
 :  68199
VITLAB® Forceps, POM, BrandTech
  83009-013
 :  Brandtech
VITLAB® Forceps, POM, BrandTech
VITLAB® Forceps, POM, BrandTech
145 mm (5¾")
$55.92
Pack of 5
Polyoxymethylene (POM) forceps with rounded ends.
  • Rounded ends
  • Opaque orange-yellow
POM is suitable for autoclaving but not suitable for microwaves. Frequent autoclaving may reduce mechanical stability. Resistant to aliphatic alcohols, ether, aldehydes, aliphatic hydrocarbons, aromatic hydrocarbons, halogenated hydrocarbons, ketones, and alkalis.
Operating temperature: –40 to 130°C (–40 to 266°F)
 : Not recommended for use with ester, acids, and oxidizing agents.
  • Longueur:
    145 mm (5¾")