VITLAB® Measuring Scoops, Polypropylene, BrandTech
50 ml (1.7 oz.), 16 cm (6¼")
Catalog # 83008-688
Supplier: Brandtech
: 39594
Jump to:
Product Details & Documents
One-piece, molded measuring scoops
- Polypropylene
- Hgih quality
Features approximate volumes marked on handle.
Spécifications
- Capacité:50 ml (1.7 oz.)
- Longueur:16 cm (6¼")